ENA429359384 Enamine

1-(5-amino-2-fluorophenyl)imidazolidin-2-one

  • MDL number MFCD11167815
  • Cas Number : 1038247-35-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-187732

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-187732
Availability details for
Quantity
  • Availability
  • Add to Cart