ENA429359385 Enamine

(3R,4S)-1-(2,2-difluoroethyl)pyrrolidine-3,4-diol

  • MDL number MFCD21919590
  • Cas Number : 1690112-93-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-187733

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-187733
Availability details for
Quantity
  • Availability
  • Add to Cart