ENA429359388 Enamine

(3-chlorothiophen-2-yl)methanol

  • MDL number MFCD06203004
  • Cas Number : 193602-41-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-187736

Safety Information

msds https://www.enaminestore.com/catalog/EN300-187736
Availability details for
Quantity
  • Availability
  • Add to Cart