ENA429359390 Enamine

N,3,3-trimethylcyclobutan-1-amine hydrochloride

  • MDL number MFCD28118665
  • Cas Number : 1803572-39-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-187738

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-187738
Availability details for
Quantity
  • Availability
  • Add to Cart