ENA429359392 Enamine

7-amino-1,2,3,4-tetrahydroquinoline-1-carboxamide

  • MDL number MFCD16702209
  • Cas Number : 1250297-08-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-187741

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-187741
Availability details for
Quantity
  • Availability
  • Add to Cart