ENA429359395 Enamine

1-(2-fluoroethyl)pyrrolidin-3-ol

  • MDL number MFCD23971971
  • Cas Number : 1556837-13-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-187744

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-187744
Availability details for
Quantity
  • Availability
  • Add to Cart