ENA429359397 Enamine

4-[2-(pyridin-4-yl)ethenyl]phenol

  • MDL number MFCD00491580
  • Cas Number : 31067-32-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-187748

Safety Information

msds https://www.enaminestore.com/catalog/EN300-187748
Availability details for
Quantity
  • Availability
  • Add to Cart