ENA429359398 Enamine

2,3,4,5-tetrahydro-1-benzoxepin-3-one

  • MDL number MFCD18450459
  • Cas Number : 35783-10-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-187749

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-187749
Availability details for
Quantity
  • Availability
  • Add to Cart