ENA429359402 Enamine

N,N-dimethyl-3-(piperidin-3-yl)-1H-pyrazole-4-carboxamide dihydrochloride

  • MDL number MFCD28118667
  • Cas Number : 1803584-06-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-187767

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-187767
Availability details for
Quantity
  • Availability
  • Add to Cart