ENA429359403 Enamine

2-(thietan-3-yl)ethan-1-ol

  • MDL number MFCD20486587
  • Cas Number : 1393688-70-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-187768

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-187768
Availability details for
Quantity
  • Availability
  • Add to Cart