ENA429359405 Enamine

4-(cyclobutoxymethyl)-3-fluorobenzonitrile

  • MDL number MFCD16684479
  • Cas Number : 1249177-89-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-187778

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-187778
Availability details for
Quantity
  • Availability
  • Add to Cart