ENA429359412 Enamine

2-[(2-{2-[(2-hydroxyethyl)sulfanyl]ethoxy}ethyl)sulfanyl]ethan-1-ol

  • MDL number MFCD00054495
  • Cas Number : 7426-02-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-187795

Safety Information

msds https://www.enaminestore.com/catalog/EN300-187795
Availability details for
Quantity
  • Availability
  • Add to Cart