ENA429359415 Enamine

1-cyclopropylpiperidine-4-carboxylic acid hydrochloride

  • MDL number MFCD08543838
  • Cas Number : 1788624-39-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-187799

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-187799
Availability details for
Quantity
  • Availability
  • Add to Cart