ENA429359417 Enamine

3-[(3-bromophenyl)methyl]-5H,6H,7H,8H-[1,2,4]triazolo[4,3-a]pyrazine dihydrochloride

  • MDL number MFCD28118670
  • Cas Number : 1798729-90-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-187801

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-187801
Availability details for
Quantity
  • Availability
  • Add to Cart