ENA429359419 Enamine

3-bromo-1-methyl-1H-pyrazole-5-carbaldehyde

  • MDL number MFCD22062891
  • Cas Number : 1367843-92-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-187803

Safety Information

msds https://www.enaminestore.com/catalog/EN300-187803
Availability details for
Quantity
  • Availability
  • Add to Cart