ENA429359421 Enamine

3-chloro-2-ethoxybenzaldehyde

  • MDL number MFCD24678078
  • Cas Number : 709649-70-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-187806

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-187806
Availability details for
Quantity
  • Availability
  • Add to Cart