ENA429359425 Enamine

2-[1-(4-fluorophenyl)-1H-1,2,3-triazol-4-yl]ethan-1-ol

  • MDL number MFCD16747680
  • Cas Number : 1249882-55-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-187813

Safety Information

msds https://www.enaminestore.com/catalog/EN300-187813
Availability details for
Quantity
  • Availability
  • Add to Cart