ENA429359426 Enamine

4-fluorobicyclo[4.2.0]octa-1(6),2,4-triene-7-carbonitrile

  • MDL number MFCD18827079
  • Cas Number : 103447-28-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-187820

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-187820
Availability details for
Quantity
  • Availability
  • Add to Cart