ENA429359430 Enamine

5-fluoro-3-imino-2,3-dihydro-1lambda6,2-benzothiazole-1,1-dione

  • MDL number MFCD28118673
  • Cas Number : 1803566-47-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-187832

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-187832
Availability details for
Quantity
  • Availability
  • Add to Cart