ENA429359431 Enamine

2,3-diamino-N,N-dimethylbenzene-1-sulfonamide

  • MDL number MFCD22371566
  • Cas Number : 1378255-41-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-187846

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-187846
Availability details for
Quantity
  • Availability
  • Add to Cart