ENA429359434 Enamine

2,3-dichloropyridine-4-carbaldehyde

  • MDL number MFCD08277282
  • Cas Number : 884495-41-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-187849

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-187849
Availability details for
Quantity
  • Availability
  • Add to Cart