ENA429359435 Enamine

1-(1-methylpiperidin-4-yl)-1H-pyrazol-3-amine

  • MDL number MFCD22393285
  • Cas Number : 1246550-66-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-187850

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-187850
Availability details for
Quantity
  • Availability
  • Add to Cart