ENA429359436 Enamine

3-(methylsulfanyl)propane-1,2-diol

  • MDL number MFCD00004724
  • Cas Number : 22551-26-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-187851

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-187851
Availability details for
Quantity
  • Availability
  • Add to Cart