ENA429359437 Enamine

1-(2-methyloxolan-2-yl)ethan-1-ol

  • MDL number MFCD23846322
  • Cas Number : 1542877-64-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-187853

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-187853
Availability details for
Quantity
  • Availability
  • Add to Cart