ENA429359439 Enamine

2-{[(2-methoxyethyl)carbamoyl]amino}-2-oxoethyl 2,3,4,5,6-pentamethylbenzoate

  • MDL number MFCD08179847
  • Cas Number : 1012212-34-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-187855

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-187855
Availability details for
Quantity
  • Availability
  • Add to Cart