ENA429359440 Enamine

4-ethoxy-3-fluorobenzene-1-carbothioamide

  • MDL number MFCD12110542
  • Cas Number : 1153271-13-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-187861

Safety Information

msds https://www.enaminestore.com/catalog/EN300-187861
Availability details for
Quantity
  • Availability
  • Add to Cart