ENA429359443 Enamine

1-benzyl-3-(thiophen-2-yl)-1H-pyrazol-5-amine

  • MDL number MFCD11558552
  • Cas Number : 222160-56-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-187864

Safety Information

msds https://www.enaminestore.com/catalog/EN300-187864
Availability details for
Quantity
  • Availability
  • Add to Cart