ENA429359446 Enamine

4-[(2-fluorophenyl)methyl]piperidin-4-ol

  • MDL number MFCD16307607
  • Cas Number : 191327-96-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-187867

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-187867
Availability details for
Quantity
  • Availability
  • Add to Cart