ENA429359461 Enamine

4-fluoro-3-(pyrrolidin-1-yl)benzoic acid hydrochloride

  • MDL number MFCD28118675
  • Cas Number : 1795284-07-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-187892

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-187892
Availability details for
Quantity
  • Availability
  • Add to Cart