ENA429359465 Enamine

ethyl 2-(aminomethyl)pentanoate hydrochloride

  • MDL number MFCD28118678
  • Cas Number : 1803566-51-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-187901

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-187901
Availability details for
Quantity
  • Availability
  • Add to Cart