ENA429359467 Enamine

1-(chloromethyl)-4-cyclobutylbenzene

  • MDL number MFCD19440006
  • Cas Number : 1379078-16-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-187904

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-187904
Availability details for
Quantity
  • Availability
  • Add to Cart