ENA429359468 Enamine

6-[(2,2,2-trifluoroethyl)amino]pyridine-3-carboxylic acid

  • MDL number MFCD11132208
  • Cas Number : 1019388-10-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-187905

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-187905
Availability details for
Quantity
  • Availability
  • Add to Cart