ENA429359469 Enamine

5-(bromomethyl)-2,4-dichloro-1,3-thiazole

  • MDL number MFCD24160467
  • Cas Number : 358672-75-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-187906

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-187906
Availability details for
Quantity
  • Availability
  • Add to Cart