ENA429359472 Enamine

2-methyl-1-oxo-1,2,3,4-tetrahydroisoquinoline-4-carboxylic acid

  • MDL number MFCD12030978
  • Cas Number : 1134335-04-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-187914

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-187914
Availability details for
Quantity
  • Availability
  • Add to Cart