ENA429359473 Enamine

5H,6H,7H,8H-imidazo[1,5-a]pyridin-7-amine dihydrochloride

  • MDL number MFCD26593755
  • Cas Number : 1417635-70-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-187915

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-187915
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart