ENA429359475 Enamine

2-fluoro-4-(furan-2-yl)aniline

  • MDL number MFCD06802088
  • Cas Number : 893736-98-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-187917

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-187917
Availability details for
Quantity
  • Availability
  • Add to Cart