ENA429359477 Enamine

ethyl 2,2-difluoro-2-(furan-2-yl)acetate

  • MDL number MFCD18071507
  • Cas Number : 698378-91-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-187928

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-187928
Availability details for
Quantity
  • Availability
  • Add to Cart