ENA429359479 Enamine

4-hydroxy-1-methyl-1H-indazole-6-carboxylic acid

  • MDL number MFCD28118680
  • Cas Number : 1556216-36-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-187938

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-187938
Availability details for
Quantity
  • Availability
  • Add to Cart