ENA429359482 Enamine

1-(3-amino-4-methylphenyl)-3-ethylurea

  • MDL number MFCD11213441
  • Cas Number : 1094458-58-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-187967

Safety Information

msds https://www.enaminestore.com/catalog/EN300-187967
Availability details for
Quantity
  • Availability
  • Add to Cart