ENA429359484 Enamine

3,3-dimethylhexanoic acid

  • MDL number MFCD02258567
  • Cas Number : 90808-83-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-187971

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-187971
Availability details for
Quantity
  • Availability
  • Add to Cart