ENA429359485 Enamine

3-amino-2-[(3,4-difluorophenyl)methyl]propanamide

  • MDL number MFCD11200908
  • Cas Number : 1038293-42-3
  • Molecular Weight : 214.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-187972

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-187972
Availability details for
Quantity
  • Availability
  • Add to Cart