ENA429359488 Enamine

3,7-dimethyl-2-(propan-2-yl)imidazo[1,2-a]pyridine

  • MDL number MFCD24199865
  • Cas Number : 1550404-29-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-187979

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-187979
Availability details for
Quantity
  • Availability
  • Add to Cart