ENA429359490 Enamine

N1-(2,2,2-trifluoroethyl)benzene-1,3-diamine

  • MDL number MFCD11101557
  • Cas Number : 1152588-11-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-187985

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-187985
Availability details for
Quantity
  • Availability
  • Add to Cart