ENA429359491 Enamine

1-[3-(1-aminoethyl)phenyl]imidazolidin-2-one

  • MDL number MFCD21213628
  • Cas Number : 1495679-23-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-187992

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-187992
Availability details for
Quantity
  • Availability
  • Add to Cart