ENA429359492 Enamine

3-[5-(trifluoromethyl)-1,2,4-oxadiazol-3-yl]aniline hydrochloride

  • MDL number MFCD16615315
  • Cas Number : 1803599-82-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-187996

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-187996
Availability details for
Quantity
  • Availability
  • Add to Cart