ENA429359498 Enamine

3,4-diethoxybenzene-1-carbothioamide

  • MDL number MFCD08164124
  • Cas Number : 60759-00-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-188005

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-188005
Availability details for
Quantity
  • Availability
  • Add to Cart