ENA429359499 Enamine

thiolane-3-carbaldehyde

  • MDL number MFCD19668858
  • Cas Number : 1065183-51-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-188006

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-188006
pictograms GHS05,GHS08
Availability details for
Quantity
  • Availability
  • Add to Cart