ENA429359500 Enamine

5-(2-fluorophenyl)-1,2-oxazol-3-amine

  • MDL number MFCD21717235
  • Cas Number : 1509591-28-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-188007

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-188007
Availability details for
Quantity
  • Availability
  • Add to Cart