ENA429359507 Enamine

3-methyl-3-phenylmorpholine

  • MDL number MFCD09901670
  • Cas Number : 933689-07-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-188015

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-188015
Availability details for
Quantity
  • Availability
  • Add to Cart