ENA429359508 Enamine

bicyclo[4.2.0]octa-1(6),2,4-trien-7-amine hydrochloride

  • MDL number MFCD22055664
  • Cas Number : 2299-00-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-188020

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-188020
Availability details for
Quantity
  • Availability
  • Add to Cart